Beginner Soldering Kit, Solar Powered LED Rocket
7505-30 7605-30
470233-462EA 23 USD
470233-462 470233-460
Beginner Soldering Kit, Solar Powered LED Rocket
The Lumen Rocket soldering kit gives beginners experience with soldering, LEDs, and solar panels.
  • Only 4 components to solder onto the rocket circuit board
  • Students intuitively learn the relationship between solar power and storage
  • Includes 18” nylon cord for hanging
  • Perfect for students 13 and up
The Rocket is a compact way for students to learn soldering. Basic instructions included, available with Red or Black circuit board. Once assembled the LED will charge either under solar power or using the standard sized USB tab on the bottom. Solar charge time is only 15 minutes in full sun, USB tab charge time is 2 minutes. Once charged the LED will gradually dim over 90 minutes. Recommended tools: soldering iron, solder, hot glue gun, hot glue, and patience.