Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

ACE® Excel C18-AR Ultra-Inert 3 and 5 micron Analytical UHPLC Compatible Columns, USP - L1, Mac Mod

Supplier: MAC-MOD ANALYTICAL MS
EXL1191546U EXL1191502U EXL1291046U EXL1291002U EXL1290503U EXL1190246U EXL1190546U EXL1190202U EXL1190502U EXL1190303U EXL1293003U EXL1193503U EXL1291503U EXL1191203U EXL1297546U EXL1297502U EXL1293503U EXL1291003U EXL1190203U EXL1292503U EXL1290346U EXL1290302U EXL1197546U EXL1197502U EXL1293046U EXL1293002U EXL1191246U EXL1191202U EXL1291246U EXL1291202U EXL1293502U EXL1293546U EXL1292502U EXL1292546U EXL1290303U EXL1290202U EXL1197503U EXL1290246U EXL1191003U EXL1191503U EXL1291203U EXL1291546U EXL1291502U EXL1290546U EXL1290502U EXL1190503U EXL1190302U EXL1190346U EXL1290203U EXL1191046U EXL1191002U EXL1193546U EXL1193502U EXL1297503U
ACE® Excel columns are are with advantage of low dispersion, ultra-high pressure UPLC and UHPLC instruments. ACE C18-AR leverages the selectivity of both C18 and Phenyl in a single phase to separate mixtures. Compatible with highly aqueous mobile phases.

  • These columns use a proprietary bonding and end-capping process to produce a very rugged and robust octadecyl-phenyl (C18-AR) phase
  • Extremely robust even when operated at high pressures and temperatures
  • Packed at extreme pressures in specialized hardware
  • Operated at pressures as high as 1,000 bar
  • A specially engineered inlet frit makes them less prone to plugging than conventional UHPLC columns

The ultra-inert characteristics of the ACE® Excel C18-AR columns make them the ideal choice for separating analytes that differ in hydrophobicity, electron-poor analytes with electron delocalization and electron withdrawing groups (i.e

ACE® Excel C18-AR has a surface area of 300 m2/g and a carbon load of 15.5%. ACE® Excel C18-AR has a recommended pH range of 2 to 8 (working outside these ranges will reduce column lifetime).

halogens, nitro groups, ketones, esters, and acids). Recommended applications for basic analytes, heterocycles, proton donors and acceptors, and halogenated aromatics. ACE® columns consistently produce measurably better peak shape and column efficiency when separating troublesome compounds. ACE® Excel C18-AR is also compatible with 100% Aqueous mobile phases for enhanced retention in reversed phase mode.



 

ORDER

Expand All / Collapse All
Column Format Phase I.D. x L Particle Size Pore Size Supplier No. VWR Catalog Number Unit Price Quantity
Analytical C18-AR 2.1 x 20 mm 3 µm 100 Å EXL1190202U 101718-606 Each Retrieving Restricted
Analytical C18-AR 3 x 20 mm 3 µm 100 Å EXL1190203U 101718-608 Each Retrieving Restricted
Analytical C18-AR 4.6 x 20 mm 3 µm 100 Å EXL1190246U 101718-610 Each Retrieving Restricted
Analytical C18-AR 2.1 x 30 mm 3 µm 100 Å EXL1190302U 101718-612 Each Retrieving Restricted
Analytical C18-AR 3 x 30 mm 3 µm 100 Å EXL1190303U 101718-614 Each Retrieving Restricted
Analytical C18-AR 4.6 x 30 mm 3 µm 100 Å EXL1190346U 101718-616 Each Retrieving Restricted
Analytical C18-AR 2.1 x 35 mm 3 µm 100 Å EXL1193502U 101718-642 Each Retrieving Restricted
Analytical C18-AR 3 x 35 mm 3 µm 100 Å EXL1193503U 101718-644 Each Retrieving Restricted
Analytical C18-AR 4.6 x 35 mm 3 µm 100 Å EXL1193546U 101718-646 Each Retrieving Restricted
Analytical C18-AR 2.1 x 50 mm 3 µm 100 Å EXL1190502U 101718-618 Each Retrieving Restricted
Analytical C18-AR 3 x 50 mm 3 µm 100 Å EXL1190503U 101718-620 Each Retrieving Restricted
Analytical C18-AR 4.6 x 50 mm 3 µm 100 Å EXL1190546U 101718-622 Each Retrieving Restricted
Analytical C18-AR 2.1 x 75 mm 3 µm 100 Å EXL1197502U 101718-648 Each Retrieving Restricted
Analytical C18-AR 3 x 75 mm 3 µm 100 Å EXL1197503U 101718-650 Each Retrieving Restricted
Analytical C18-AR 4.6 x 75 mm 3 µm 100 Å EXL1197546U 101718-652 Each Retrieving Restricted
Analytical C18-AR 2.1 x 100 mm 3 µm 100 Å EXL1191002U 101718-624 Each Retrieving Restricted
Analytical C18-AR 3 x 100 mm 3 µm 100 Å EXL1191003U 101718-626 Each Retrieving Restricted
Analytical C18-AR 4.6 x 100 mm 3 µm 100 Å EXL1191046U 101718-628 Each Retrieving Restricted
Analytical C18-AR 2.1 x 125 mm 3 µm 100 Å EXL1191202U 101718-630 Each Retrieving Restricted
Analytical C18-AR 3 x 125 mm 3 µm 100 Å EXL1191203U 101718-632 Each Retrieving Restricted
Analytical C18-AR 4.6 x 125 mm 3 µm 100 Å EXL1191246U 101718-634 Each Retrieving Restricted
Analytical C18-AR 2.1 x 150 mm 3 µm 100 Å EXL1191502U 101718-636 Each Retrieving Restricted
Analytical C18-AR 3 x 150 mm 3 µm 100 Å EXL1191503U 101718-638 Each Retrieving Restricted
Analytical C18-AR 4.6 x 150 mm 3 µm 100 Å EXL1191546U 101718-640 Each Retrieving Restricted
Analytical C18-AR 2.1 x 20 mm 5 µm 100 Å EXL1290202U 101719-074 Each Retrieving Restricted
Analytical C18-AR 3 x 20 mm 5 µm 100 Å EXL1290203U 101719-076 Each Retrieving Restricted
Analytical C18-AR 4.6 x 20 mm 5 µm 100 Å EXL1290246U 101719-078 Each Retrieving Restricted
Analytical C18-AR 2.1 x 30 mm 5 µm 100 Å EXL1290302U 101719-080 Each Retrieving Restricted
Analytical C18-AR 3 x 30 mm 5 µm 100 Å EXL1290303U 101719-082 Each Retrieving Restricted
Analytical C18-AR 4.6 x 30 mm 5 µm 100 Å EXL1290346U 101719-084 Each Retrieving Restricted
Analytical C18-AR 2.1 x 35 mm 5 µm 100 Å EXL1293502U 101719-122 Each Retrieving Restricted
Analytical C18-AR 3 x 35 mm 5 µm 100 Å EXL1293503U 101719-124 Each Retrieving Restricted
Analytical C18-AR 4.6 x 35 mm 5 µm 100 Å EXL1293546U 101719-126 Each Retrieving Restricted
Analytical C18-AR 2.1 x 50 mm 5 µm 100 Å EXL1290502U 101719-086 Each Retrieving Restricted
Analytical C18-AR 3 x 50 mm 5 µm 100 Å EXL1290503U 101719-088 Each Retrieving Restricted
Analytical C18-AR 4.6 x 50 mm 5 µm 100 Å EXL1290546U 101719-090 Each Retrieving Restricted
Analytical C18-AR 2.1 x 75 mm 5 µm 100 Å EXL1297502U 101719-128 Each Retrieving Restricted
Analytical C18-AR 3 x 75 mm 5 µm 100 Å EXL1297503U 101719-130 Each Retrieving Restricted
Analytical C18-AR 4.6 x 75 mm 5 µm 100 Å EXL1297546U 101719-132 Each Retrieving Restricted
Analytical C18-AR 2.1 x 100 mm 5 µm 100 Å EXL1291002U 101719-092 Each Retrieving Restricted
Analytical C18-AR 3 x 100 mm 5 µm 100 Å EXL1291003U 101719-094 Each Retrieving Restricted
Analytical C18-AR 4.6 x 100 mm 5 µm 100 Å EXL1291046U 101719-096 Each Retrieving Restricted
Analytical C18-AR 2.1 x 125 mm 5 µm 100 Å EXL1291202U 101719-098 Each Retrieving Restricted
Analytical C18-AR 3 x 125 mm 5 µm 100 Å EXL1291203U 101719-100 Each Retrieving Restricted
Analytical C18-AR 4.6 x 125 mm 5 µm 100 Å EXL1291246U 101719-102 Each Retrieving Restricted
Analytical C18-AR 2.1 x 150 mm 5 µm 100 Å EXL1291502U 101719-104 Each Retrieving Restricted
Analytical C18-AR 3 x 150 mm 5 µm 100 Å EXL1291503U 101719-106 Each Retrieving Restricted
Analytical C18-AR 4.6 x 150 mm 5 µm 100 Å EXL1291546U 101719-108 Each Retrieving Restricted
Analytical C18-AR 2.1 x 250 mm 5 µm 100 Å EXL1292502U 101719-110 Each Retrieving Restricted
Analytical C18-AR 3 x 250 mm 5 µm 100 Å EXL1292503U 101719-112 Each Retrieving Restricted
Analytical C18-AR 4.6 x 250 mm 5 µm 100 Å EXL1292546U 101719-114 Each Retrieving Restricted
Analytical C18-AR 2.1 x 300 mm 5 µm 100 Å EXL1293002U 101719-116 Each Retrieving Restricted
Analytical C18-AR 3 x 300 mm 5 µm 100 Å EXL1293003U 101719-118 Each Retrieving Restricted
Analytical C18-AR 4.6 x 300 mm 5 µm 100 Å EXL1293046U 101719-120 Each Retrieving Restricted
restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty