Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Chiral Columns, Regis Phase, ES Industries

Supplier: ES INDUSTRIES
780201 780101 786615
The Whelk-O 1 is useful for the separation of underivatized enantiomers in a number of families including amides, epoxides, esters, ureas, carbamates, ethers, aziridines, phosphonates, aldehydes, ketones, carboxylic acids, alcohols and non-steroidal anti-inflammatory drugs (NSAIDs).

  • Covalent bonding ensures universal solvent compatibility and excellent durability
  • Fast method development and run times
  • Allows for the inversion of peak elution order
  • Broad versatility comparable to polysaccharide-derived chiral stationary phases
  • Columns are packed and tested at high pressures for use in both SFC and HPLC mode

This π-electron acceptor/π-electron donor phase exhibits an extraordinary degree of generality. The broad versatility observed on the Whelk-O 1 column compares favorably with polysaccharide-derived chiral stationary phases.




 

ORDER

Expand All / Collapse All
Column Format Phase I.D. x L Particle Size Supplier No. VWR Catalog Number Unit Price Quantity
Analytical (R,R)-Whelk-O®1 4.6 x 250 mm 5 µm 780201 97015-586 Each Retrieving
Analytical (S,S)-Whelk-O®1 4.6 x 250 mm 5 µm 780101 97015-584 Each Retrieving
Analytical (S,S)-Whelk-O®1 4.6 x 250 mm 10 µm 786615 89235-172 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty