Anti-Mitochondrial fission regulator 1 Mouse Polyclonal Antibody

Supplier: Novus Biologicals
H00009650-B01P
103337-436EA 798.46 USD
103337-436
Anti-Mitochondrial fission regulator 1 Mouse Polyclonal Antibody
Antibodies
The Mitochondrial fission regulator 1 Antibody from Novus Biologicals is a mouse polyclonal antibody to Mitochondrial fission regulator 1. This antibody reacts with human. The Mitochondrial fission regulator 1 Antibody has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: Mitochondrial fission regulator 1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR