Anti-Mitochondria Mouse Monoclonal Antibody [clone: MTC719]

Supplier: Novus Biologicals
NBP2-45218
103282-724EA 515.69 USD
103282-724
Anti-Mitochondria Mouse Monoclonal Antibody [clone: MTC719]
Antibodies
The Mitochondria Antibody (MTC719) - IHC-Prediluted from Novus Biologicals is a mouse monoclonal antibody to Mitochondria. This antibody reacts with human. The Mitochondria Antibody (MTC719) - IHC-Prediluted has been validated for the following applications: Immunohistochemistry-Paraffin.

Type: Primary
Antigen: Mitochondria
Clonality: Monoclonal
Clone: MTC719
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR