Anti-Methyltransferase like 5 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-92107
103277-040EA 734.58 USD
103277-040
Anti-Methyltransferase like 5 Rabbit Polyclonal Antibody
Antibodies
The Methyltransferase like 5 Antibody from Novus Biologicals is a rabbit polyclonal antibody to Methyltransferase like 5. This antibody reacts with human. The Methyltransferase like 5 Antibody has been validated for the following applications: Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: Methyltransferase like 5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR