Anti-Methionine Sulfoxide Reductase B Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
H00051734-D01P
103344-442EA 844.62 USD
103344-442
Anti-Methionine Sulfoxide Reductase B Rabbit Polyclonal Antibody
Antibodies
The Methionine Sulfoxide Reductase B Antibody from Novus Biologicals is a rabbit polyclonal antibody to Methionine Sulfoxide Reductase B. This antibody reacts with human, mouse. The Methionine Sulfoxide Reductase B Antibody has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: Methionine Sulfoxide Reductase B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR