Anti-Methionine Sulfoxide Reductase A Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
H00004482-D01P
103331-120EA 844.62 USD
103331-120
Anti-Methionine Sulfoxide Reductase A Rabbit Polyclonal Antibody
Antibodies
The Methionine Sulfoxide Reductase A Antibody from Novus Biologicals is a rabbit polyclonal antibody to Methionine Sulfoxide Reductase A. This antibody reacts with human, mouse. The Methionine Sulfoxide Reductase A Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: Methionine Sulfoxide Reductase A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR