Anti-Meteorin-like/METRNL Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-91064
103279-042EA 734.58 USD
103279-042
Anti-Meteorin-like/METRNL Rabbit Polyclonal Antibody
Antibodies
The Meteorin-like / METRNL Antibody from Novus Biologicals is a rabbit polyclonal antibody to Meteorin-like / METRNL. This antibody reacts with human. The Meteorin-like / METRNL Antibody has been validated for the following applications: Immunohistochemistry, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: Meteorin-like/METRNL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR