Anti-Metallothionein-3 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-89772
103277-252EA 777.05 USD
103277-252
Anti-Metallothionein-3 Rabbit Polyclonal Antibody
Antibodies
The Metallothionein-3 Antibody from Novus Biologicals is a rabbit polyclonal antibody to Metallothionein-3. This antibody reacts with human. The Metallothionein-3 Antibody has been validated for the following applications: Immunohistochemistry, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: Metallothionein-3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR