Anti-Mesothelin Mouse Monoclonal Antibody [clone: 1A10]

Supplier: Novus Biologicals
H00010232-M02
103338-476EA 767.69 USD
103338-476
Anti-Mesothelin Mouse Monoclonal Antibody [clone: 1A10]
Antibodies
The Mesothelin Antibody (1A10) from Novus Biologicals is a mouse monoclonal antibody to Mesothelin. This antibody reacts with human. The Mesothelin Antibody (1A10) has been validated for the following applications: Western Blot, ELISA, Immunoprecipitation.

Type: Primary
Antigen: Mesothelin
Clonality: Monoclonal
Clone: 1A10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR