Anti-Menin Mouse Polyclonal Antibody

Supplier: Novus Biologicals
H00004221-B01P
103330-780EA 399 USD
103330-780
Anti-Menin Mouse Polyclonal Antibody
Antibodies
The Menin Antibody from Novus Biologicals is a mouse polyclonal antibody to Menin. This antibody reacts with human. The Menin Antibody has been validated for the following applications: Western Blot, Immunocytochemistry / Immunofluorescence.

Type: Primary
Antigen: Menin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR