Anti-Melanotransferrin/CD228/MFI2 Mouse Polyclonal Antibody

Supplier: Novus Biologicals
H00004241-B01P
103330-832EA 399 USD
103330-832
Anti-Melanotransferrin/CD228/MFI2 Mouse Polyclonal Antibody
Antibodies
The Melanotransferrin / CD228 / MFI2 Antibody from Novus Biologicals is a mouse polyclonal antibody to Melanotransferrin / CD228 / MFI2. This antibody reacts with human. The Melanotransferrin / CD228 / MFI2 Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: Melanotransferrin/CD228/MFI2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR