Anti-Mediator of cell motility 1 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-88269
103271-552EA 734.58 USD
103271-552
Anti-Mediator of cell motility 1 Rabbit Polyclonal Antibody
Antibodies
The mediator of cell motility 1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to mediator of cell motility 1. This antibody reacts with human, mouse, rat. The mediator of cell motility 1 Antibody has been validated for the following applications: Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: Mediator of cell motility 1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR