Anti-Matrilin-2 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
H00004147-D01P
103330-546EA 429 USD
103330-546
Anti-Matrilin-2 Rabbit Polyclonal Antibody
Antibodies
The Matrilin-2 Antibody from Novus Biologicals is a rabbit polyclonal antibody to Matrilin-2. This antibody reacts with human. The Matrilin-2 Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: Matrilin-2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR