Anti-Marapsin/Pancreasin Mouse Monoclonal Antibody [clone: 6F11]

Supplier: Novus Biologicals
H00083886-M02
103344-962EA 798.46 USD
103344-962
Anti-Marapsin/Pancreasin Mouse Monoclonal Antibody [clone: 6F11]
Antibodies
The Marapsin / Pancreasin Antibody (6F11) from Novus Biologicals is a mouse monoclonal antibody to Marapsin / Pancreasin. This antibody reacts with human. The Marapsin / Pancreasin Antibody (6F11) has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: Marapsin/Pancreasin
Clonality: Monoclonal
Clone: 6F11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR