Anti-MAP3K12 binding inhibitory protein 1 Mouse Monoclonal Antibody [clone: 200]

Supplier: Novus Biologicals
H00051562-M05
103344-290EA 798.46 USD
103344-290
Anti-MAP3K12 binding inhibitory protein 1 Mouse Monoclonal Antibody [clone: 200]
Antibodies
The MAP3K12 binding inhibitory protein 1 Antibody (2E2) from Novus Biologicals is a mouse monoclonal antibody to MAP3K12 binding inhibitory protein 1. This antibody reacts with human. The MAP3K12 binding inhibitory protein 1 Antibody (2E2) has been validated for the following applications: ELISA, Immunocytochemistry / Immunofluorescence.

Type: Primary
Antigen: MAP3K12 binding inhibitory protein 1
Clonality: Monoclonal
Clone: 200
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR