Anti-MAD2L1-binding protein Mouse Monoclonal Antibody [clone: 4G11]

Supplier: Novus Biologicals
H00009587-M03
103337-318EA 798.46 USD
103337-318
Anti-MAD2L1-binding protein Mouse Monoclonal Antibody [clone: 4G11]
Antibodies
The MAD2L1-binding protein Antibody (4G11) from Novus Biologicals is a mouse monoclonal antibody to MAD2L1-binding protein. This antibody reacts with human. The MAD2L1-binding protein Antibody (4G11) has been validated for the following applications: Western Blot, ELISA, Immunocytochemistry / Immunofluorescence.

Type: Primary
Antigen: MAD2L1-binding protein
Clonality: Monoclonal
Clone: 4G11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR