Anti-Lysyl Oxidase Homolog 3/LOXL3 Rabbit Polyclonal Antibody

NBP1-85908
103273-676EA 607.69 USD
103273-676
Anti-Lysyl Oxidase Homolog 3/LOXL3 Rabbit Polyclonal Antibody
The Lysyl Oxidase Homolog 3 / LOXL3 Antibody from Novus Biologicals is a rabbit polyclonal antibody to Lysyl Oxidase Homolog 3 / LOXL3. This antibody reacts with human. The Lysyl Oxidase Homolog 3 / LOXL3 Antibody has been validated for the following applications: Immunohistochemistry, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: Lysyl Oxidase Homolog 3/LOXL3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR