Anti-Lysosomal Pro-X Carboxypeptidase/PRCP Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
H00005547-D01P
103308-010EA 844.62 USD
103308-010
Anti-Lysosomal Pro-X Carboxypeptidase/PRCP Rabbit Polyclonal Antibody
Antibodies
The Lysosomal Pro-X Carboxypeptidase / PRCP Antibody from Novus Biologicals is a rabbit polyclonal antibody to Lysosomal Pro-X Carboxypeptidase / PRCP. This antibody reacts with human. The Lysosomal Pro-X Carboxypeptidase / PRCP Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: Lysosomal Pro-X Carboxypeptidase/PRCP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR