Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Lunatic Fringe Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
H00003955-D01P
103330-134
Anti-Lunatic Fringe Rabbit Polyclonal Antibody
The Lunatic Fringe Antibody from Novus Biologicals is a rabbit polyclonal antibody to Lunatic Fringe. This antibody reacts with human. The Lunatic Fringe Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: Lunatic Fringe
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now


 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty