Anti-Luciferase Rabbit Polyclonal Antibody

NBP2-19296
102709-464EA 493.85 USD
102709-464
Anti-Luciferase Rabbit Polyclonal Antibody
The Luciferase Antibody from Novus Biologicals is a rabbit polyclonal antibody to Luciferase. This antibody reacts with firefly. The Luciferase Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: Luciferase
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Firefly
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR