Anti-Luciferase Goat Polyclonal Antibody

Supplier: Novus Biologicals
NB100-1677SS
103368-500EA 281.66 USD
103368-500
Anti-Luciferase Goat Polyclonal Antibody
Antibodies
The Luciferase Antibody from Novus Biologicals is a goat polyclonal antibody to Luciferase. This antibody reacts with firefly. The Luciferase Antibody has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Type: Primary
Antigen: Luciferase
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Firefly
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR