Anti-Livin Mouse Monoclonal Antibody (DyLight 755) [clone: 88C570]

Supplier: Novus Biologicals
NB100-56548IR
103360-084EA 614.15 USD
103360-084
Anti-Livin Mouse Monoclonal Antibody (DyLight 755) [clone: 88C570]
Antibodies
The Livin Antibody (88C570) [DyLight 755] from Novus Biologicals is a mouse monoclonal antibody to Livin. This antibody reacts with human. The Livin Antibody (88C570) [DyLight 755] has been validated for the following applications: Western Blot, Immunocytochemistry / Immunofluorescence.

Type: Primary
Antigen: Livin
Clonality: Monoclonal
Clone: 88C570
Conjugation: DyLight 755
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR