Anti-Liprin alpha 1 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
21720002-0.1MG
103317-684EA 399 USD
103317-684
Anti-Liprin alpha 1 Rabbit Polyclonal Antibody
Antibodies
The Liprin alpha 1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to Liprin alpha 1. This antibody reacts with human. The Liprin alpha 1 Antibody has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Type: Primary Antibodies
Antigen: Liprin alpha 1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR