Anti-Lewis A Blood Group Antigen Mouse Polyclonal Antibody

Supplier: Novus Biologicals
H00002525-B01P
103326-832EA 399 USD
103326-832
Anti-Lewis A Blood Group Antigen Mouse Polyclonal Antibody
Antibodies
The Lewis A Blood Group Antigen Antibody from Novus Biologicals is a mouse polyclonal antibody to Lewis A Blood Group Antigen. This antibody reacts with human. The Lewis A Blood Group Antigen Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: Lewis A Blood Group Antigen
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR