Anti-Leukotriene A4 Hydrolase/LTA4H Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-90375
103277-808EA 734.58 USD
103277-808
Anti-Leukotriene A4 Hydrolase/LTA4H Rabbit Polyclonal Antibody
Antibodies
The Leukotriene A4 Hydrolase / LTA4H Antibody from Novus Biologicals is a rabbit polyclonal antibody to Leukotriene A4 Hydrolase / LTA4H. This antibody reacts with human. The Leukotriene A4 Hydrolase / LTA4H Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: Leukotriene A4 Hydrolase/LTA4H
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR