Anti-Leucyl-cystinyl Aminopeptidase/LNPEP Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP2-32111
103287-420EA 635.51 USD
103287-420
Anti-Leucyl-cystinyl Aminopeptidase/LNPEP Rabbit Polyclonal Antibody
Antibodies
The Leucyl-cystinyl Aminopeptidase / LNPEP Antibody from Novus Biologicals is a rabbit polyclonal antibody to Leucyl-cystinyl Aminopeptidase / LNPEP. This antibody reacts with human. The Leucyl-cystinyl Aminopeptidase / LNPEP Antibody has been validated for the following applications: Western Blot, Immunoprecipitation.

Type: Primary
Antigen: Leucyl-cystinyl Aminopeptidase/LNPEP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR