Anti-Legumain/Asparaginyl Endopeptidase Mouse Monoclonal Antibody [clone: M1]

Supplier: Novus Biologicals
H00005641-M02
103308-376EA 798.46 USD
103308-376
Anti-Legumain/Asparaginyl Endopeptidase Mouse Monoclonal Antibody [clone: M1]
Antibodies
The Legumain / Asparaginyl Endopeptidase Antibody (M1) from Novus Biologicals is a mouse monoclonal antibody to Legumain / Asparaginyl Endopeptidase. This antibody reacts with human. The Legumain / Asparaginyl Endopeptidase Antibody (M1) has been validated for the following applications: ELISA.

Type: Primary
Antigen: Legumain/Asparaginyl Endopeptidase
Clonality: Monoclonal
Clone: M1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR