Anti-Legumain/Asparaginyl Endopeptidase Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
H00005641-D01P
103308-374EA 429 USD
103308-374
Anti-Legumain/Asparaginyl Endopeptidase Rabbit Polyclonal Antibody
Antibodies
The Legumain / Asparaginyl Endopeptidase Antibody from Novus Biologicals is a rabbit polyclonal antibody to Legumain / Asparaginyl Endopeptidase. This antibody reacts with human. The Legumain / Asparaginyl Endopeptidase Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: Legumain/Asparaginyl Endopeptidase
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR