Anti-Latent TGF-beta bp4 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP2-43671
103123-546EA 635.51 USD
103123-546
Anti-Latent TGF-beta bp4 Rabbit Polyclonal Antibody
Antibodies
The Latent TGF-beta bp4 Antibody from Novus Biologicals is a rabbit polyclonal antibody to Latent TGF-beta bp4. This antibody reacts with human, mouse. The Latent TGF-beta bp4 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: Latent TGF-beta bp4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR