Anti-Latent TGF-beta bp2/LTBP-2 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-88411
103274-802EA 734.58 USD
103274-802
Anti-Latent TGF-beta bp2/LTBP-2 Rabbit Polyclonal Antibody
Antibodies
The Latent TGF-beta bp2 / LTBP-2 Antibody from Novus Biologicals is a rabbit polyclonal antibody to Latent TGF-beta bp2 / LTBP-2. This antibody reacts with human. The Latent TGF-beta bp2 / LTBP-2 Antibody has been validated for the following applications: Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: Latent TGF-beta bp2/LTBP-2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR