Anti-Latent TGF-beta bp1 Mouse Polyclonal Antibody

H00004052-A01
103330-250EA 436.92 USD
103330-250
Anti-Latent TGF-beta bp1 Mouse Polyclonal Antibody
The Latent TGF-beta bp1 Antibody from Novus Biologicals is a mouse polyclonal antibody to Latent TGF-beta bp1. This antibody reacts with human. The Latent TGF-beta bp1 Antibody has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: Latent TGF-beta bp1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR