Anti-Lactalbumin Alpha Mouse Monoclonal Antibody (DyLight 755) [clone: F20.16]

NB100-2783IR
103357-616EA 466.15 USD
103357-616
Anti-Lactalbumin Alpha Mouse Monoclonal Antibody (DyLight 755) [clone: F20.16]
The Lactalbumin Alpha Antibody (F20.16) [DyLight 755] from Novus Biologicals is a mouse monoclonal antibody to Lactalbumin Alpha. This antibody reacts with human. The Lactalbumin Alpha Antibody (F20.16) [DyLight 755] has been validated for the following applications: Western Blot, ELISA, Immunocytochemistry / Immunofluorescence.

Type: Primary
Antigen: Lactalbumin Alpha
Clonality: Monoclonal
Clone: F20.16
Conjugation: DyLight 755
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR