Anti-Kinesin C2 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP2-17061
102709-060EA 635.51 USD
102709-060
Anti-Kinesin C2 Rabbit Polyclonal Antibody
Antibodies
The Kinesin C2 Antibody from Novus Biologicals is a rabbit polyclonal antibody to Kinesin C2. This antibody reacts with human, mouse. The Kinesin C2 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: Kinesin C2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR