Anti-Kinesin Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP2-21667
102713-354EA 777.05 USD
102713-354
Anti-Kinesin Rabbit Polyclonal Antibody
Antibodies
The Kinesin Antibody from Novus Biologicals is a rabbit polyclonal antibody to Kinesin. This antibody reacts with human. The Kinesin Antibody has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry.

Type: Primary
Antigen: Kinesin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR