Zinc chloride, anhydrous 99.99% (trace metals basis)
|
Danger
|
Synonyms:
Zinc (II) chloride
, Butter of zinc
AC38013-0250
AC38013-0050
103528-128EA
244.46
USD
103528-128
103528-126
Zinc chloride, anhydrous 99.99% (trace metals basis)
Zinc chloride
Formula:
ZnCl₂ MW: 136.3 g/mol Boiling Pt: 732 °C (760 mmHg) Melting Pt: 293 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011295 CAS Number: 7646-85-7 EINECS: 231-592-0 UN: 2331 ADR: 8,III Merck Index: 12,10261 |
Specification Test Results
Appearance | White to off-white Crystalline powder or crystals |
Total trace metal impurities | ≤0.01 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...