Xylenol orange tetrasodium salt ACS

Synonyms: Xylenol orange water soluble

AC44760-0050
103518-122EA 83.9 USD
103518-122
Xylenol orange tetrasodium salt ACS
Xylenol orange tetrasodium salt
Formula: C₃₁H₂₈N₂Na₄O₁₃S
MW: 760.59 g/mol
Melting Pt: 210 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00044293
CAS Number: 3618-43-7

Order Now

Specification Test Results

Appearance Red-brown to dark brown Crystalline powder
Suitability (for Zn titration) Passes test
Clarity of solution Passes test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR