Xylene (mixture of isomers), Extra Pure

Danger

Synonyms: Dimethylbenzene

AC39079-0010 AC39079-0100 AC39079-0025
103528-426EA 240.2 USD
103528-426 103528-424 103528-422
Xylene (mixture of isomers), Extra Pure
Xylene (mixture of isomers)
Formula: C₆H₄(CH₃)₂
MW: 106.17 g/mol
Boiling Pt: 136…140 °C (760 mmHg)
Melting Pt: –34 °C
Density: 0.865 g/cm³
Flash Pt: 23 °C
Storage Temperature: Ambient
MDL Number: MFCD00077264
CAS Number: 1330-20-7
EINECS: 215-535-7
UN: 1307
ADR: 3,III
Merck Index: 12,10214
https://us.vwr.com/stibo/web/std.lang.all/44/47/704447.jpg

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Water ≤0.05 %
GC ≥98.0 % (mixed isomers)
Residue after evaporation ≤0.001 %
Substance darkened by H2SO4 Passes test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR