Xylene (mixture of isomers) 98+% for analysis

Danger

Synonyms: Dimethylbenzene

AC38394-0010 AC38394-0050
103528-224EA 295.38 USD
103528-224 103528-222
Xylene (mixture of isomers) 98+% for analysis
Xylene (mixture of isomers)
Formula: C₆H₄(CH₃)₂
MW: 106.17 g/mol
Boiling Pt: 136…140 °C (760 mmHg)
Melting Pt: –34 °C
Density: 0.865 g/cm³
Flash Pt: 23 °C
Storage Temperature: Ambient
MDL Number: MFCD00077264
CAS Number: 1330-20-7
EINECS: 215-535-7
UN: 1307
ADR: 3,III
Merck Index: 12,10214
https://us.vwr.com/stibo/web/std.lang.all/44/47/704447.jpg

Order Now

Specification Test Results

Appearance Clear liquid
Water ≤0.05 % (K.F.)
GC ≥98.0 % (isomers and ethylbenzene)
Color scale ≤10 APHA
Residue after evaporation ≤0.002 %
Acidity/alkalinity ≤0.0001 meq/g
Calcium (Ca) ≤0.5 ppm
Copper (Cu) ≤0.05 ppm
Iron (Fe) ≤0.1 ppm
Lead (Pb) ≤0.05 ppm
Magnesium (Mg) ≤0.2 ppm
Potassium (K) ≤0.5 ppm
Sodium (Na) ≤0.5 ppm
Zinc (Zn) ≤0.1 ppm
Impurity ≤1 % Toluene
Impurity ≤1 % n-Propyl benzene
Benzene ≤0.05 %
KMnO4-reducing substance ≤5 ppm (O)
Total phosphorus (P) ≤0.1 ppm
Total silicon (Si) ≤0.05 ppm
Total sulfur (S) ≤2 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR