Water, Extra Pure, Deionized

Synonyms: Dihydrogen oxide

AC34547-0050 AC34547-0250
103466-872EA 59.54 USD
103466-872 103464-438
Water, Extra Pure, Deionized
Water
Formula: H₂O
MW: 18.02 g/mol
Boiling Pt: 100 °C (760 mmHg)
Density: 1 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011332
CAS Number: 7732-18-5
EINECS: 231-791-2
Merck Index: 12,10175

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Residue after evaporation ≤25 ppm
Conductivity ≤5.0 µS/cm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR