Water for HPLC

Synonyms: Dihydrogen oxide

AC38939-0010 AC38939-0025
103467-242EA 97.74 USD
103467-242 103467-240
Water for HPLC
Water
Formula: H₂O
MW: 18.02 g/mol
Boiling Pt: 100 °C (760 mmHg)
Density: 1 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011332
CAS Number: 7732-18-5

Order Now

Specification Test Results

Appearance Clear colorless Liquid
UV at 200 nm A: ≤0.05
UV at 210 nm A: ≤0.009
UV at 254 nm A: ≤0.004
Residue after evaporation ≤0.0003 %
Conductivity ≤1 µS/cm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR