Tris acetate 99%
Synonyms:
Tris(hydroxymethyl)aminomethane acetate
, 2-Amino-2-(hydroxymethyl)-1,3-propanediol acetate
, Trimethylolaminomethane acetate
, 2-Amino-2-(hydroxymethyl)propane-1,3-diol acetate salt
, Tris(hydroxymethyl)aminomethane acetate salt
, Tris acetate
AC43450-1000
AC43450-5000
103467-200EA
146
USD
103467-200
103467-202
Tris acetate 99%
Tris acetate
Formula:
NH₂C(CH₂OH)₃·CH₃COOH MW: 181.1870 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00038951 CAS Number: 6850-28-8 |
Specification Test Results
Appearance | White Crystalline powder |
Water | ≤1.0 % |
Solubility | (1 g/ml in water) Clear colorless |
UV | (50 % in water) |
UV | at 290 nm A: ≤0.5 |
Titration with NaOH | ≥98.5 % |
Heavy metals (as Pb) | ≤5 ppm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...