Trimethylsulfonium hydroxide 0.25 M in methanol

Danger

Synonyms: TMSH

AC38136-0010 AC38136-0100
103528-154EA 341.54 USD
103528-154 103528-152
Trimethylsulfonium hydroxide 0.25 M in methanol
Trimethylsulfonium Hydroxide
Formula: C₃H₁₀OS
MW: 94.17 g/mol
Density: 0.81 g/cm³
Flash Pt: 11 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00216756
CAS Number: 17287-03-5
UN: 3286
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless Solution
Titration with HCl 0.20 to 0.30 M

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR