Triisopropyl phosphite 96%

Danger

Synonyms: Tri-i-propylphosphite

AC37238-0050 AC37238-0025 AC37238-1000
103527-998EA 521 USD
103527-998 103528-002 103528-000
Triisopropyl phosphite 96%
Triisopropyl phosphite
Formula: C₉H₂₁O₃P
MW: 208.24 g/mol
Boiling Pt: 94…95 °C (66.6hPa)
Density: 0.914 g/cm³
Flash Pt: 62 °C
Storage Temperature: Ambient
MDL Number: MFCD00008874
CAS Number: 116-17-6
EINECS: 204-130-0
UN: 3278
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4095 to 1.4115 (20 °C, 589 nm)
GC ≥95 %
Color scale ≤25 APHA

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR