Trifluorothymidine 98%

Warning

Synonyms: Trifluoromethyldeoxyuridine5-Trifluorothymidine1-((2R,4S,5R)-4-Hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-5-(trifluoromethyl)pyrimidine-2,4(1H,3H)-dione

AC45291-2500
103466-702EA 191 USD
103466-702
Trifluorothymidine 98%
Trifluorothymidine
Formula: C₁₀H₁₁F₃N₂O₅
MW: 296.2 g/mol
Melting Pt: 190…193 °C
Storage Temperature: Freezer
MDL Number: MFCD00006534
CAS Number: 70-00-8

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR