Trifluoroacetic acid 99+% for spectroscopy

Danger

Synonyms: TFA

AC43416-1000
103524-402EA 195 USD
103524-402
Trifluoroacetic acid 99+% for spectroscopy
Trifluoroacetic acid
Formula: C₂HF₃O₂
MW: 114.02 g/mol
Boiling Pt: 72 °C (760 mmHg)
Melting Pt: –15 °C
Density: 1.535 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00004169
CAS Number: 76-05-1
UN: 2699
ADR: 8,I
https://us.vwr.com/stibo/web/std.lang.all/83/92/728392.jpg

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Water ≤0.05 %
UV at 320 nm A: ≤0.03
UV at 300 nm A: ≤0.03
UV at 290 nm A: ≤0.04
UV at 280 nm A: ≤0.05
UV at 270 nm A: ≤0.10
UV at 260 nm A: ≤1.0
Color scale ≤10 APHA
Residue after evaporation ≤0.005 %
Titration with NaOH ≥99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR