Triethyloxonium tetrafluoroborate 95%

Danger

AC43676-0250 AC43676-1000
103528-866EA 238 USD
103528-866 103528-864
Triethyloxonium tetrafluoroborate 95%
Triethyloxonium tetrafluoroborate
Formula: C₆H₁₅BF₄O
Melting Pt: 84…89 °C
Storage Temperature: Freezer
MDL Number: MFCD00044423
CAS Number: 368-39-8
UN: 2925
ADR: 4.1,II

Order Now

Specification Test Results

Appearance White to off-white Powder
Stabilizer 3 to 5 % ether
Titration with NaOH ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR