Triethylamine 99.5% for analysis

Danger

Synonyms: TEA(Diethylamino)ethane

AC43228-0010 AC43228-0025
103528-794EA 268 USD
103528-794 103524-396
Triethylamine 99.5% for analysis
Triethylamine
Formula: (C₂H₅)₃N
MW: 101.19 g/mol
Boiling Pt: 90 °C (760 mmHg)
Melting Pt: –115 °C
Density: 0.728 g/cm³
Flash Pt: –11 °C
Storage Temperature: Ambient
MDL Number: MFCD00009051
CAS Number: 121-44-8
UN: 1296
ADR: 3,II
https://us.vwr.com/stibo/web/std.lang.all/92/82/729282.jpg

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.3990 to 1.4010 (20 °C, 589 nm)
Water ≤0.1 %
GC ≥99.4 %
GC ≤0.05 % (diethylamine)
GC ≤0.05 % (ethanol)
Color scale ≤15 APHA
Residue after evaporation ≤0.002 %
Chloride (Cl) ≤10 ppm
Sulfate (SO4) ≤10 ppm
Cadmium (Cd) ≤0.05 ppm
Calcium (Ca) ≤0.5 ppm
Chromium (Cr) ≤0.02 ppm
Cobalt (Co) ≤0.02 ppm
Copper (Cu) ≤0.02 ppm
Iron (Fe) ≤0.1 ppm
Lead (Pb) ≤0.1 ppm
Magnesium (Mg) ≤0.1 ppm
Manganese (Mn) ≤0.02 ppm
Nickel (Ni) ≤0.02 ppm
Zinc (Zn) ≤0.1 ppm
Heavy metals (as Pb) ≤1 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR