Tributyl borate 98%, AcroSeal™

Warning

Synonyms: Tri-n-butylborate

AC42866-8000 AC42866-1000
103466-674EA 266 USD
103466-674 103466-668
Tributyl borate 98%, AcroSeal™
Tributyl borate
Formula: C₁₂H₂₇BO₃
MW: 230.15 g/mol
Boiling Pt: 230…235 °C (760 mmHg)
Melting Pt: –70 °C
Density: 0.853 g/cm³
Flash Pt: 93 °C
Storage Temperature: Ambient
MDL Number: MFCD00009434
CAS Number: 688-74-4

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4080 to 1.4100 (20 °C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR